missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CSF2RA / GM-CSFR / CD116 Protein

A DNA sequence encoding the Homo sapiens (Human) CSF2RA / GM-CSFR / CD116, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5877-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958389

  • 2285.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001155001.1
CSF2RA / GM-CSFR / CD116 Protein
Research Use Only
CSF2RA
Recombinant Protein
E. coli
EKSDLRTVAP ASSLNVRFDS RTMNLSWDCQ ENTTFSKCFL TDKKNRVVEP RLSNNECSCT FREICLHEGV TFEVHVNTSQ RGFQQKLLYP NSGREGTAAQ NFSCFIYNAD LMNCTWARGP TAPRDVQYFL YIRNSKRRRE IRCPYYIQDS GTHVGCHLDN LSGLTSRNYF LVNGTSREIG IQFFDSLLDT KKIERFNPPS NVTVRCNTTH CLVRWKQPRT YQKLSYLDFQ YQLDVHRKNT QPGTENLLIN VSGDLENRYN FPSSEPRAKH SVKIRAADVR ILNWSSWSEA IEFGSDDG
Greater than 90% as determined by SDS-PAGE.
1438
1 mg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers