missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CSNK2A2 Protein
A cDNA sequence encoding the CSNK2A2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11533-2ug
Additional Details : Weight : 0.01000kg
Specifications
1459 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMPGPAA GSRARVYAEV NSLRSREYWD YEAHVPSWGN QDDYQLVRKL GRGKYSEVFE AINITNNERV VVKILKPVKK KKIKREVKIL ENLRGGTNII KLIDTVKDPV SKTPALVFEY INNTDFKQLY QILTDFDIRF YMYELLKALD YCHSKGIMHR DVKPHNVMID HQQKKLRLID WGLAEFYHPA QEYNVRVASR YFKGPELLVD YQMYDYSLDM WSLGCMLASM IFRREPFFHG QDNYDQLVRI AKVLGTEELY GYLKKYHIDL DPHFNDILGQ HSRKRWENFI HSENRHLVSP EALDLLDKLL RYDHQQRLTA KEAMEHPYFY PVVKEQSQPC ADNAVLSSGL TAAR | |
Greater than 90% as determined by SDS-PAGE. |
CSNK2A2 Protein | |
Research Use Only | |
CSNK2A2 | |
Human | |
His | |
CSNK2A2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT. |