missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CST1 Protein
A cDNA sequence encoding the CST1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11535-1mg
Additional Details : Weight : 0.01000kg
Specifications
CST1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMWSPKEE DRIIPGGIYN ADLNDEWVQR ALHFAISEYN KATKDDYYRR PLRVLRARQQ TVGGVNYFFD VEVGRTICTK SQPNLDTCAF HEQPELQKKQ LCSFEIYEVP WENRRSLVKS RCQES | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CST1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 10% glycerol and 100mM NaCl. |