missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CTLA4 Protein
A cDNA sequence encoding the CTLA4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11557-HIS-1mg
Specifications
1493 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADLKAMHVAQ PAVVLASSRG IASFVCEYAS PGKATEVRVT VLRQADSQVT EVCAATYMMG NELTFLDDSI CTGTSSGNQV NLTIQGLRAM DTGLYICKVE LMYPPPYYLG IGNGTQIYVI DPEPCPDSDH HHHHH | |
Greater than 90% as determined by SDS-PAGE. |
CTLA4 Protein | |
Research Use Only | |
CTLA4 | |
Human | |
His | |
CTLA4 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 20% glycerol. |