missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CTSZ Protein

A cDNA sequence encoding the CTSZ was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11564-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927658

  • 3915.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1522
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSLPKSWDW RNVDGVNYAS ITRNQHIPQY CGSCWAHAST SAMADRINIK RKGAWPSTLL SVQNVIDCGN AGSCEGGNDL SVWDYAHQHG IPDETCNNYQ AKDQECDKFN QCGTCNEFKE CHAIRNYTLW RVGDYGSLSG REKMMAEIYA NGPISCGIMA TERLANYTGG IYAEYQDTTY INHVVSVAGW GISDGTEYWI VRNSWGEPWG ERGWLRIVTS TYKDGKGARY NLAIEEHCTF GDPIVNormal0falsefalsefalseEN-USX-NONEHE-->
Greater than 90.0% as determined by SDS-PAGE.
CTSZ Protein
Research Use Only
CTSZ
Human
His
The CTSZ solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers