missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CUL4A/RBX1/DDB1/CRBN Complex Protein

Product Code. 18715846
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18715846

Brand: R&D Systems™ E3650025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Typical concentration to support in vitro conjugation reactions will depend on experimental conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13619.3, P62877.1, Q16531.1, Q96SW2.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl, DTT and Glycerol.
Molecular Weight (g/mol) M.W. Theroretical: 94 kDa (CUL4A), 12 kDa (RB x 1), 127 kDa (DDB1), 51 kDa (CRBN)
Quantity 25 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human CUL4A/RB x 1/DDB1/CRBN Complex protein This complex consists of an N-terminal 10-His tagged Cullin-4 A, untagged RB x 1, untagged DDB1 and untagged CRBN.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Regulatory Status RUO
Specific Reactivity CUL4A/RBX1/DDB1/CRBN Complex
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer X mg/mL (X μm) in 50 mM HEPES pH 7.5, 200 mM NaCl, 10% (v/v) Glycerol, 1 mM DTT
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein CUL4A/RBX1/DDB1/CRBN Complex
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.