missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CUZD1 Protein
A cDNA sequence encoding the CUZD1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | CUZD1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15927678
|
enQuireBio™
QP11570-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15907678
|
enQuireBio™
QP11570-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15917678
|
enQuireBio™
QP11570-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CUZD1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CUZD1 was filtered (0.4microm) and lyophilized in phosphate buffered saline and 5% (w/v) trehalose. |
Research Use Only | |
Recombinant Protein | |
Human (HEK293) | |
EAEGNASCTV SLGGANMAET HKAMILQLNP SENCTWTIER PENKSIRIIF SYVQLDPDGS CESENIKVFD GTSSNGPLLG QVCSKNDYVP VFESSSSTLT FQIVTDSARI QRTVFVFYYF FSPNISIPNC GGYLDTLEGS FTSPNYPKPH PELAYCVWHI QVEKDYKIKL NFKEIFLEID KQCKFDFLAI YDGPSTNSGL IGQVCGRVTP TFESSSNSLT VVLSTDYANS YRGFSASYTS IYAENINTTS LTCSSDRMRV IISKSYLEAF NSNGNNLQLK DPTCRPKLSN VVEFSVPLNG CGTIRKVEDQ SITYTNIITF SASSTSEVIT RQKQLQIIVK CEMGHNSTVE IIYITEDDVI QSQNALGKYN TSMALFESNS FEKTILESPY YVDLNQTLFV QVSLHTSDPN LVVFLDTCRA SPTSDFASPT YDLIKSGCSR DETCKVYPLF GHYGRFQFNA FKFLRSMSSV YLQCKVLICD SSDHQSRCNQ GCVSRSKRDI SSYKWKTDSI IGPIRLKRDR SASGNSGFQH ETHAEETPNQ PFNS HHHHHH HHHH | |
Greater than 95.0% as determined by SDS-PAGE. |