missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL13 / BCA-1 / BLC Protein
Recombinant Protein
Brand: enQuireBio™ QP911-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006410.1 | |
CXCL13 / BCA-1 / BLC Protein | |
Research Use Only | |
CXCL13 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
10563 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
VLEVYYTSLR . CRVQESSVF . IPRRFIDRI Q. ILPRGNGC PR. KEIIVWK KNK. SIVCVD PQAE. WIQRM MEVLR. KRSS STLPVP. VFK RKIP | |
Greater than 95% as determined by SDS-PAGE. |