missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CYB5R3 Protein
A cDNA sequence encoding the CYB5R3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11575-20ug
This item is not returnable.
View return policy
Specifications
CYB5R3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMFQRST PAITLESPDI KYPLRLIDRE IISHDTRRFR FALPSPQHIL GLPVGQHIYL SARIDGNLVV RPYTPISSDD DKGFVDLVIK VYFKDTHPKF PAGGKMSQYL ESMQIGDTIE FRGPSGLLVY QGKGKFAIRP DKKSNPIIRT VKSVGMIAGG TGITPMLQVI RAIMKDPDDH TVCHLLFANQ TEKDILLRPE LEELRNKHSA RFKLWYTLDR APEAWDYGQG FVNEEMIRDH LPPPEEEPLV LMCGPPPMIQ YACLPNLDHV GHPTERCFVF | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CYB5R3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 10% glycerol. |