missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cyclophilin G Protein
A cDNA sequence encoding the Cyclophilin G was constructed and used to recombinantly synthesize the protein.
131.00 € - 3275.00 €
Specifications
Gene ID (Entrez) | 9360 |
---|---|
Name | Cyclophilin G Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | PPIG |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15904298
|
enQuireBio™
QP10574-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15994288
|
enQuireBio™
QP10574-25UG |
25 μg |
203.00 €
25µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984288
|
enQuireBio™
QP10574-1MG |
1 mg |
3275.00 €
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
9360 | |
Research Use Only | |
PPIG | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGIKVQRPRC FFDIAINNQP AGRVVFELFS DVCPKTCENF RCLCTGEKGT GKSTQKPLHY KSCLFHRVVK DFMVQGGDFS EGNGRGGESI YGGFFEDESF AVKHNKEFLL SMANRGKDTN GSQFFITTKP TPHLDGHHVV FGQVISGQEV VREIENQKTD AASKPFAEVR ILSCG | |
Greater than 95.0% as determined by SDS-PAGE. |
Cyclophilin G Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 200 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin. | |
Human | |
His | |
Cyclophilin-G solution containing 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol. |