missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cysteine and glycine-rich protein 1
A DNA sequence encoding the Homo sapiens (Human) Cysteine and glycine-rich protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8561-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001287894.1 | |
Cysteine and glycine-rich protein 1 | |
Research Use Only | |
CSRP2 | |
Recombinant Protein | |
E. coli | |
PVWGGGNKCG ACGRTVYHAE EVQCDGRSFH RCCFLCMVCR KNLDSTTVAI HDEEIYCKSC YGKKYGPKGY GYGQGAGTLN MDRGERLGIK PESVQPHRPT TNPNTSKFAQ KYGGAEKCSR CGDSVYAAEK IIGAGKPWHK NCFRCAKCGK SLESTTLTEK EGEIYCKGCY AKNFGPKGFG YGQGAGALVH AQ | |
Greater than 90% as determined by SDS-PAGE. |
1466 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |