missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CYTH2 Protein
A cDNA sequence encoding the CYTH2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11590-20ug
Additional Details : Weight : 0.01000kg
Specifications
CYTH2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEDGVYE PPDLTPEERM ELENIRRRKQ ELLVEIQRLR EELSEAMSEV EGLEANEGSK TLQRNRKMAM GRKKFNMDPK KGIQFLVENE LLQNTPEEIA RFLYKGEGLN KTAIGDYLGE REELNLAVLH AFVDLHEFTD LNLVQALRQF LWSFRLPGEA QKIDRMMEAF AQRYCLCNPG VFQSTDTCYV LSFAVIMLNT SLHNPNVRDK PGLERFVAMN RGINEGGDLP EELLRNLYDS IRNEPFKIPE DDGNDLTHTF FNPDREGWLL KLGGRVKTWK RRWFILTDNC LYYFEYTTDK EPRGIIPLEN LSIREVDDPR KPNCFELYIP NNKGQLIKAC KTEADGRVVE GNHMVYRISA PTQEEKDEWI KSIQAAVSVD PFYEMLAARK KRISVKKKQE QP | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CYTH2 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 1mM DTT and 10% glycerol. |