missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DBR1 Protein

A cDNA sequence encoding the DBR1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11598-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957758

  • 203.00 € / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

DBR1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHVKFAAL MQHQAKDKGQ TARATKFLAL DKCLPHRDFL QILEIEHDPS APDYLEYDIE WLTILRATDD LINVTGRLWN MPENNGLHAR WDYSATEEGM KEVLEKLNHD LKVPCNFSVT AACYDPSKPQ TQMQLIHRIN PQTTEFCAQL GIIDINVRLQ KSKEEHHVCG EYEEQDDVES NDSGEDQSEY NTDTSALSSI NPDEIMLDEE EDEDSIVSAH SGMNTPSVEP SDQASEFSAS FSDVRILPGS MIVSSDDTVD STIDREGKPG GTVESGNGED LTKVPLKRLS DEHEPEQRKK IKRRNQAIYA AVDDDDDDAA
Greater than 90.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The DBR1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers