missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human DC-SIGN/CD209 Fc Chimera Protein

Product Code. 18797496
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18797496

Brand: R&D Systems™ 11265DC050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHO-Expressed

Measured by the ability of the immobilized protein to support the adhesion of ICAM-3 expressing CHO Chinese hamster ovary cells. The ED50 for this effect is 0.700-7.00 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NNX6.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Molecular Weight (g/mol) M.W.-observed: 62-74 kDa, under reducing conditions., M.W.-theroretical: 65.7 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human DC-SIGN/CD209 protein MD Human IgG1 (Pro100-Lys330) IEGR Human DC-SIGN/CD209 (Lys62-Ala404) (C-terminus) Accession # Q9NNX6.1 (N-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.