missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DDX56 Protein

A cDNA sequence encoding the DDX56 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11614-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957808

  • 131.00 € / 5µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

DDX56 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMEDSEAL GFEHMGLDPR LLQAVTDLGW SRPTLIQEKA IPLALEGKDL LARARTGSGK TAAYAIPMLQ LLLHRKATGP VVEQAVRGLV LVPTKELARQ AQSMIQQLAT YCARDVRVAN VSAAEDSVSQ RAVLMEKPDV VVGTPSRILS HLQQDSLKLR DSLELLVVDE ADLLFSFGFE EELKSLLCHL PRIYQAFLMS ATFNEDVQAL KELILHNPVT LKLQESQLPG PDQLQQFQVV CETEEDKFLL LYALLKLSLI RGKSLLFVNT LERSYRLRLF LEQFSIPTCV LNGELPLRSR CHIISQFNQG FYDCVIATDA EVLGAPVKGK RRGRGPKGDK ASDPEAGVAR GIDFHHVSAV LNFDLPPTPE AYIHRAGRTA RANNPGIVLT FVLPTEQFHL GKIEELLSGE NRGPILLPYQ FRMEEIEGFR YRCRDAMRSV TKQAIREARL KEIKEELLHS EKLKTYFEDN PRDLQLLRHD LPLHPAVVKP HLGHVPDYLV PPALRGLVRP HKKRKKLSSS CRKAKRAKSQ NPLRSFKHKG KKFRPTAKPS
Greater than 85.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
DDX56 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers