missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dermcidin / DCD Protein

A DNA sequence encoding the Homo sapiens (Human) Dermcidin / DCD, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5914-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958639

  • 615.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001287783.1
Dermcidin / DCD Protein
Research Use Only
DCD
Recombinant Protein
Yeast
YDPEAASAPG SGNPCHEASA AQKENAGEDP GLARQAPKPR KQRSSLLEKG LDGAKKAVGG LGKLGKDAVE DLESVGKGAV HDVKDVLDSV L
Greater than 90% as determined by SDS-PAGE.
117159
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers