missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dermcidin / DCD Protein
A DNA sequence encoding the Homo sapiens (Human) Dermcidin / DCD, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5914-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001287783.1 | |
Dermcidin / DCD Protein | |
Research Use Only | |
DCD | |
Recombinant Protein | |
Yeast | |
YDPEAASAPG SGNPCHEASA AQKENAGEDP GLARQAPKPR KQRSSLLEKG LDGAKKAVGG LGKLGKDAVE DLESVGKGAV HDVKDVLDSV L | |
Greater than 90% as determined by SDS-PAGE. |
117159 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |