missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Desmoglein-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Desmoglein-3, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001935.2 |
---|---|
Gene ID (Entrez) | 1830 |
Name | Desmoglein-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15928879
|
enQuireBio™
QP5948-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948879
|
enQuireBio™
QP5948-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938879
|
enQuireBio™
QP5948-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15958879
|
enQuireBio™
QP5948-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15968879
|
enQuireBio™
QP5948-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15978879
|
enQuireBio™
QP5948-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001935.2 | |
Desmoglein-3 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
1830 | |
Research Use Only | |
Dsg3 | |
Recombinant Protein | |
E. coli | |
EWVKFAKPCR EGEDNSKRNP IAKITSDYQA TQKITYRISG VGIDQPPFGI FVVDKNTGDI NITAIVDREE TPSFLITCRA LNAQGLDVEK PLILTVKILD INDNPPVFSQ QIFMGEIEEN SASNSLVMIL NATDADEPNH LNSKIAFKIV SQEPAGTPMF LLSRNTGEVR TLTNSLDREQ ASSYRLVVSG ADKDGEGLST QCECNIKVKD VNDNFPMFRD SQYSARIEEN ILSSELLRFQ VTDLDEEYTD NWLAVYFFTS GNEGNWFEIQ TDPRTNEGIL KVVKALDYEQ LQSVKLSIAV KNKAEFHQSV ISRYRVQSTP VTIQVINVRE GIAFRPASKT FTVQKGISSK KLVDYILGTY QAIDEDTNKA ASNVKYVMGR NDGGYLMIDS KTAEIKFVKN MNRDSTFIVN KTITAEVLAI DEYTGKTSTG TVYVRVPDFN DNCPTAVLEK DAVCSSSPSV VVSARTLNNR YTGPYTFALE DQPVKLPAVW SITTLNATSA LLRAQEQIPP GVYHISLVLT DSQNNRCEMP RSLTLEVCQC DNRGICGTSY PTTSPGTRYG RPHSGR | |
Greater than 90% as determined by SDS-PAGE. |