missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DHRS2 Protein

A DNA sequence encoding the Homo sapiens (Human) DHRS2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7548-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049320

  • 3260.00 € / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005785.1
DHRS2 Protein
Research Use Only
DHRS2
Recombinant Protein
E. coli
MLSAVARGYQ GWFHPCARLS VRMSSTGIDR KGVLANRVAV VTGSTSGIGF AIARRLARDG AHVVISSRKQ QNVDRAMAKL QGEGLSVAGI VCHVGKAEDR EQLVAKALEH CGGVDFLVCS AGVNPLVGST LGTSEQIWDK ILSVNVKSPA LLLSQLLPYM ENRRGAVILV SSIAAYNPVV ALGVYNVSKT ALLGLTRTLA LELAPKDIRV NCVVPGIIKT DFSKVVRIGF MGMSLSGRTS RNIISCRGLG SQRTVQESCP SCALQMPATS TGRTLRWQAT PLGSERSGGG CVAVVPGPGA
Greater than 80% as determined by SDS-PAGE.
10202
1 mg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers