missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DHRS4 Protein

A cDNA sequence encoding the DHRS4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11665-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927968

  • 181.00 € / 5µg
Estimated Shipment: 19-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

DHRS4 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The DHRS4 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 20% glycerol and 1mM DTT.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMHKAGL LGLCARAWNS VRMASSGMTR RDPLANKVAL VTASTDGIGF AIARRLAQDG AHVVVSSRKQ QNVDQAVATL QGEGLSVTGT VCHVGKAEDR ERLVATAVKL HGGIDILVSN AAVNPFFGSI MDVTEEVWDK TLDINVKAPA LMTKAVVPEM EKRGGGSVVI VSSIAAFSPS PGFSPYNVSK TALLGLTKTL AIELAPRNIR VNCLAPGLIK TSFSRMLWMD KEKEESMKET LRIRRLGEPE DCAGIVSFLC SEDASYITGE TVVVGGGTPS RL
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers