missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Diacylglycerol kinase alpha Protein

A DNA sequence encoding the Homo sapiens (Human) Diacylglycerol kinase alpha, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5930-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958749

  • 2285.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001336.2
Diacylglycerol kinase alpha Protein
Research Use Only
DGKA
Recombinant Protein
E. coli
MAKERGLISP SDFAQLQKYM EYSTKKVSDV LKLFEDGEMA KYVQGDAIGY EGFQQFLKIY LEVDNVPRHL SLALFQSFET GHCLNETNVT KDVVCLNDVS CYFSLLEGGR PEDKLEFTFK LYDTDRNGIL DSSEVDKIIL QMMRVAEYLD WDVSELRPIL QEMMKEIDYD GSGSVSQAEW VRAGATTVPL LVLLGLEMTL KDDGQHMWRP KRFPRPVYCN LCESSIGLGK QGLSCNLCKY TVHDQCAMKA LPCEVSTYAK SRKDIGVQSH VWVRGGCESG RCDRCQKKIR IYHSLTGLHC VWCHLEIHDD CLQAVGHECD CGLLRDHILP PSSIYPSVLA SGPDRKNSKT SQKTMDDLNL STSEALRIDP VPNTHPLLVF VNPKSGGKQG QRVLWKFQYI LNPRQVFNLL KDGPEIGLRL FKDVPDSRIL VCGGDGTVGW ILETIDKANL PVLPPVAVLP LGTGNDLARC LRWGGGYEGQ NLAKILKDLE MSKVVHMDRW SVEVIPQQTE EKSDPVPFQI INNYFSIGVD ASIAHRFHIM REKYPEKFNS RMKNKLWYFE FATSESIFST CKKLEESLTV EICGKPLDLS NLSLEGIAVL NIPSMHGGSN LWGDTRRPHG DIYGINQALG ATAKVITDPD ILKTCVPDLS DKRLEVVGLE GAIEMGQIYT KLKNAGRRLA KCSEITFHTT KTLPMQIDGE PWMQTPCTIK ITHKNQMPML MGPPPRSTNF FGFLS
Greater than 80% as determined by SDS-PAGE.
1606
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers