missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DLD Protein
A cDNA sequence encoding the DLD was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11669-5ug
Additional Details : Weight : 0.01000kg
Specifications
1738 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMADQ PIDADVTVIG SGPGGYVAAI KAAQLGFKTV CIEKNETLGG TCLNVGCIPS KALLNNSHYY HMAHGKDFAS RGIEMSEVRL NLDKMMEQKS TAVKALTGGI AHLFKQNKVV HVNGYGKITG KNQVTATKAD GGTQVIDTKN ILIATGSEVT PFPGITIDED TIVSSTGALS LKKVPEKMVV IGAGVIGVEL GSVWQRLGAD VTAVEFLGHV GGVGIDMEIS KNFQRILQKQ GFKFKLNTKV TGATKKSDGK IDVSIEAASG GKAEVITCDV LLVCIGRRPF TKNLGLEELG IELDPRGRIP VNTRFQTKIP NIYAIGDVVA GPMLAHKAED EGIICVEGMA GGAVHIDYNC VPSVIYTHPE VAWVGKSEEQ LKEEGIEYKV GKFPFAANSR AKTNADTDGM VKILGQKSTD RVLGAHILGP GAGEMVNEAA LALEYGASCE DIARVCHAHP TLSEAFREAN LAASFGKSIN F | |
Greater than 95% as determined by SDS-PAGE. |
DLD Protein | |
Research Use Only | |
DLD | |
Human | |
His | |
The DLD solution contains 20mM Tris-HCl pH 8.0, 1mM DTT, 0.1M NaCl and 10% glycerol. |