missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DLD Protein

A cDNA sequence encoding the DLD was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11669-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987978

  • 3275.00 € / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1738
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMADQ PIDADVTVIG SGPGGYVAAI KAAQLGFKTV CIEKNETLGG TCLNVGCIPS KALLNNSHYY HMAHGKDFAS RGIEMSEVRL NLDKMMEQKS TAVKALTGGI AHLFKQNKVV HVNGYGKITG KNQVTATKAD GGTQVIDTKN ILIATGSEVT PFPGITIDED TIVSSTGALS LKKVPEKMVV IGAGVIGVEL GSVWQRLGAD VTAVEFLGHV GGVGIDMEIS KNFQRILQKQ GFKFKLNTKV TGATKKSDGK IDVSIEAASG GKAEVITCDV LLVCIGRRPF TKNLGLEELG IELDPRGRIP VNTRFQTKIP NIYAIGDVVA GPMLAHKAED EGIICVEGMA GGAVHIDYNC VPSVIYTHPE VAWVGKSEEQ LKEEGIEYKV GKFPFAANSR AKTNADTDGM VKILGQKSTD RVLGAHILGP GAGEMVNEAA LALEYGASCE DIARVCHAHP TLSEAFREAN LAASFGKSIN F
Greater than 95% as determined by SDS-PAGE.
DLD Protein
Research Use Only
DLD
Human
His
The DLD solution contains 20mM Tris-HCl pH 8.0, 1mM DTT, 0.1M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers