missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human DLL4 Fc Chimera Protein

Product Code. 18766976
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18766976 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18766976 Supplier R&D Systems™ Supplier No. 10185D4050
  • ONLINE EXCLUSIVE PRICE
    508.48 €
    458.85 € / 50µg
    Save 49.63 €
    9% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Delta-like protein 4 (DLL4) is a type I membrane protein belonging to the Delta/Serrate/Lag2 (DSL) family of Notch ligands

  • Notch signaling is an evolutionarily conserved pathway that controls cell fate and is required in multiple developmental processes including vascular development, hematopoiesis, somatogenesis, myogenesis, and neurogenesis
  • Dysregulation in the Notch pathway is associated with various human diseases
  • Measured by the ability of the immobilized protein to enhance BMP-2 induced alkaline phosphatase activity in C3H10T1/2 mouse embryonic fibroblast cells. The ED50 fort his effect is 0.02-0.12 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NR61
For Use With (Application) Bioactivity
Gene ID (Entrez) 54567
Molecular Weight (g/mol) 81 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human DLL4 protein

Human DLL4 (Ser27-Pro524) Accession # Q9NR61IEGRMDPHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.