missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DnaJ homolog subfamily B member 2 Protein

A DNA sequence encoding the Homo sapiens (Human) DnaJ homolog subfamily B member 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5938-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928809

  • 2095.00 € / 1mg
Estimated Shipment: 19-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001034639.1
DnaJ homolog subfamily B member 2 Protein
1 mg
Not determined.
Full Length of Isoform 3
Human
His-SUMO
Tris-based buffer, 50% glycerol
3300
Greater than 90% as determined by SDS-PAGE.
Research Use Only
DNAJB2
Recombinant Protein
E. coli
ASYYEILDVP RSASADDIKK AYRRKALQWH PDKNPDNKEF AEKKFKEVAE AYEVLSDKHK REIYDRYGRE GLTGTGTGPS RAEAGSGGPG FTFTFRSPEE VFREFFGSGD PFAELFDDLG PFSELQNRGS RHSGPFFTFS SSFPGHSDFS SSSFSFSPGA GAFRSVSTST TFVQGRRITT RRIMENGQER VEVEEDGQLK SVTINGVPDD LALGLELSRR EQQPSVTSRS GGTQVQQTPA SCPLDSDLSE DEDLQLAMAY SLSEMEAAGK KPAGGREAQH RRQGRPKAQH QDPGLGGTQE GARGEATKRS PSPEEKASRC LIL
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers