missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNase Protein
A cDNA sequence encoding the DNase was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11692-1500IU
Additional Details : Weight : 0.01000kg
Specifications
1773 | |
1500 IU | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
CHO | |
MRGMKLLGAL LALAALLQGA VSLKIAAFNI QTFGETKMSN ATLVSYIVQI LSRYDIALVQ EVRDSHLTAV GKLLDNLNQD APDTYHYVVS EPLGRNSYKE RYLFVYRPDQ VSAVDSYYYD DGCEPCGNDT FNREPAIVRF FSRFTEVREF AIVPLHAAPG DAVAEIDALY DVYLDVQEKW GLEDVMLMGD FNAGCSYVRP SQWSSIRLWT SPTFQWLIPD SADTTATPTH CAYDRIVVAG MLLRGAVVPD SALPFNFQAA YGLSDQLAQA ISDHYPVEVM LK |
DNase Protein | |
Research Use Only | |
DNASE1 | |
Human | |
Untagged | |
Each mg contains 150 microg calcium chloride dehydrate and 8.77 mg NaCl. |