missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNTTIP1 Protein
A DNA sequence encoding the Homo sapiens (Human) DNTTIP1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_443183.1 |
---|---|
Gene ID (Entrez) | 116092 |
Name | DNTTIP1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15908839
|
enQuireBio™
QP5942-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928839
|
enQuireBio™
QP5942-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15918839
|
enQuireBio™
QP5942-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15938839
|
enQuireBio™
QP5942-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15948839
|
enQuireBio™
QP5942-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15958839
|
enQuireBio™
QP5942-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_443183.1 | |
DNTTIP1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
116092 | |
Research Use Only | |
DNTTIP1 | |
Recombinant Protein | |
E. coli | |
MGATGDAEQP RGPSGAERGG LELGDAGAAG QLVLTNPWNI MIKHRQVQRR GRRSQMTTSF TDPAISMDLL RAVLQPSINE EIQTVFNKYM KFFQKAALNV RDNVGEEVDA EQLIQEACRS CLEQAKLLFS DGEKVIPRLT HELPGIKRGR QAEEECAHRG SPLPKKRKGR PPGHILSSDR AAAGMVWKPK SCEPIRREGP KWDPARLNES TTFVLGSRAN KALGMGGTRG RIYIKHPHLF KYAADPQDKH WLAEQHHMRA TGGKMAYLLI EEDIRDLAAS DDYRGCLDLK LEELKSFVLP SWMVEKMRKY METLRTENEH RAVEAPPQT | |
Greater than 90% as determined by SDS-PAGE. |