missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Docking protein 1
A DNA sequence encoding the Homo sapiens (Human) Docking protein 5, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_060901.2 |
---|---|
Gene ID (Entrez) | 55816 |
Name | Docking protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15968839
|
enQuireBio™
QP5943-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988839
|
enQuireBio™
QP5943-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978839
|
enQuireBio™
QP5943-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15998839
|
enQuireBio™
QP5943-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15908849
|
enQuireBio™
QP5943-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15918849
|
enQuireBio™
QP5943-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_060901.2 | |
Docking protein 1 | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
55816 | |
Research Use Only | |
DOK5 | |
Recombinant Protein | |
E. coli | |
MECVGTRIND ISLGEPDLLA TGVEREQSER FNVYLMPSPN LDVHGECALQ ITYEYICLWD VQNPRVKLIS WPLSALRRYG RDTTWFTFEA GRMCETGEGL FIFQTRDGEA IYQKVHSAAL AIAEQHERLL QSVKNSMLQM KMSERAASLS TMVPLPRSAY WQHITRQHST GQLYRLQDVS SPLKLHRTET FPAYRSEH | |
Greater than 90% as determined by SDS-PAGE. |