missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DPY30 Protein
A cDNA sequence encoding the DPY30 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11701-1mg
Additional Details : Weight : 0.01000kg
Specifications
DPY30 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEPEQML EGQTQVAENP HSEYGLTDNV ERIVENEKIN AEKSSKQKVD LQSLPTRAYL DQTVVPILLQ GLAVLAKERP PNPIEFLASY LLKNKAQFED RN | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
DPY30 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 30% glycerol and 2mM DTT. |