missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dynamin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Dynamin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5939-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001005336.1 | |
Dynamin-1 Protein | |
Research Use Only | |
DNM1 | |
Recombinant Protein | |
E. coli | |
GNRGMEDLIP LVNRLQDAFS AIGQNADLDL PQIAVVGGQS AGKSSVLENF VGRDFLPRGS GIVTRRPLVL QLVNATTEYA EFLHCKGKKF TDFEEVRLEI EAETDRVTGT NKGISPVPIN LRVYSPHVLN LTLVDLPGMT KVPVGDQPPD IEFQIRDMLM QFVTKENCLI LAVSPANSDL ANSDALKVAK EVDPQGQRTI GVITKLDLMD EGTDARDVLE NKLLPLRRGY IGVVNRSQKD IDGK | |
Greater than 90% as determined by SDS-PAGE. |
1759 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |