missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DYNLRB2 Protein
A cDNA sequence encoding the DYNLRB2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11719-1mg
Additional Details : Weight : 0.01000kg
Specifications
DYNLRB2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAEVEETLKR IQSHKGVIGT MVVNAEGIPI RTTLDNSTTV QYAGLLHHLT MKAKSTVRDI DPQNDLTFLR IRSKKHEIMV APDKEYLLIV IQNPCE | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The DYNLRB2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |