missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E Selectin Protein
A cDNA sequence encoding the E Selectin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11723-2ug
Additional Details : Weight : 0.01000kg
Specifications
E Selectin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MIASQFLSAL TLVLLIKESG AWSYNTSTEA MTYDEASAYC QQRYTHLVAI QNKEEIEYLN SILSYSPSYY WIGIRKVNNV WVWVGTQKPL TEEAKNWAPG EPNNRQKDED CVEIYIKREK DVGMWNDERC SKKKLALCYT AACTNTSCSG HGECVETINN YTCKCDPGFS GLKCEQIVNC TALESPEHGS LVCSHPLGNF SYNSSCSISC DRGYLPSSME TMQCMSSGEW SAPIPACNVV ECDAVTNPAN GFVECFQNPG SFPWNTTCTF DCEEGFELMG AQSLQCTSSG NWDNEKPTCK AVTCRAVRQP QNGSVRCSHS PAGEFTFKSS CNFTCEEGFM LQGPAQVECT TQGQWTQQIP VCEAFQCTAL SNPERGYMNC LPSASGSFRY GSSCEFSCEQ GFVLKGSKRL QCGPTGEWDN EKPTCEAVRC DAVHQPPKGL VRCAHSPIGE FTYKSSCAFS CEEGFELHGS TQLECTSQGQ WTEEVPSCQV VKCSSLAVPG KINMSCSGEP VFGTVCKFAC PEGWTLNGSA ARTCGATGHW SGLLPTCEAP TESNIPLVAG LSAAGLSLLT LAPFLLWLRK CLRKAKKFVP ASSCQSLESD GSYQKPSYIL | |
Greater than 95% as determined by SDS-PAGE. Single band on Western Blot. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
E-Selectin is supplied in 1x PBS and 50% Glycerol. |