missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase RFWD2 Protein

Product Code. p-7147942
Click to view available options
Quantity:
10 μg
100 μg
50 μg
Unit Size:
100µg
10µg
50µg
This item is not returnable. View return policy

Product Code. 15932748

Brand: enQuireBio™ QP10010iv10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens(Human) E3 ubiquitin-protein ligase RFWD2, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001001740.1
Gene ID (Entrez) 64326
Name E3 ubiquitin-protein ligase RFWD2 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol RFWD2
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MSGSRQAGSG SAGTSPGSSA ASSVTSASSS LSSSPSPPSV AVSAAALVSG GVAQAAGSGG LGGPVRPVLV APAVSGSGGG AVSTGLSRHS CAARPSAGVG GSSSSLGSGS RKRPLLAPLC NGLINSYEDK SNDFVCPICF DMIEEAYMTK CGHSFCYKCI HQSLEDNNRC PKCNYVVDNI DHLYPNFLVN ELILKQKQRF EEKRFKLDHS VSSTNGHRWQ IFQDWLGTDQ DNLDLANVNL MLELLVQKKK QLEAESHAAQ LQILMEFLKV ARRNKREQLE QIQKELSVLE EDIKRVEEMS GLYSPVSEDS TVPQFEAPSP SHSSIIDSTE YSQPPGFSGS SQTKKQPWYN STLASRRKRL TAHFEDLEQC YFSTRMSRIS DDSRTASQLD EFQECLSKFT RYNSVRPLAT LSYASDLYNG SSIVSSIEFD RDCDYFAIAG VTKKIKVYEY DTVIQDAVDI HYPENEMTCN SKISCISWSS YHKNLLASSD YEGTVILWDG FTGQRSKVYQ EHEKRCWSVD FNLMDPKLLA SGSDDAKVKL WSTNLDNSVA SIEAKANVCC VKFSPSSRYH LAFGCADHCV HYYDLRNTKQ PIMVFKGHRK AVSYAKFVSG EEIVSASTDS QLKLWNVGKP YCLRSFKGHI NEKNFVGLAS NGDYIACGSE NNSLYLYYKG LSKTLLTFKF DTVKSVLDKD RKEDDTNEFV SAVCWRALPD GESNVLIAAN SQGTIKVLEL V
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.