missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase SMURF2 Protein

A DNA sequence encoding the Homo sapiens (Human) E3 ubiquitin-protein ligase SMURF2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8114-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042741

  • 615.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_073576.1
E3 ubiquitin-protein ligase SMURF2 Protein
Research Use Only
SMURF2
Recombinant Protein
Yeast
MSNPGGRRNG PVKLRLTVLC AKNLVKKDFF RLPDPFAKVV VDGSGQCHST DTVKNTLDPK WNQHYDLYIG KSDSVTISVW NHKKIHKKQG AGFLGCVRLL SNAINRLKDT GYQRLDLCKL GPNDNDTVRG QIVVSLQSRD RIGTGGQVVD CSRLFDNDLP DGWEERRTAS GRIQYLNHIT RTTQWERPTR PASEYSSPGR PLSCFVDENT PISGTNGATC GQSSDPRLAE RRVRSQRHRN YMSRTHLHTP PDLPEGYEQR TTQQGQVYFL HTQTGVSTWH DPRVPRDLSN INCEELGPLP PGWEIRNTAT GRVYFVDHNN RTTQFTDPRL SANLHLVLNR QNQLKDQQQQ QVVSLCPDDT ECLTVPRYKR DLVQKLKILR QELSQQQPQA GHCRIEVSRE EIFEESYRQV MKMRPKDLWK RLMIKFRGEE GLDYGGVARE WLYLLSHEML NPYYGLFQYS RDDIYTLQIN PDSAVNPEHL SYFHFVGRIM GMAVFHGHYI DGGFTLPFYK QLLGKSITLD DMELVDPDLH NSLVWILEND ITGVLDHTFC VEHNAYGEII QHELKPNGKS IPVNEENKKE YVRLYVNWRF LRGIEAQFLA LQKGFNEVIP QHLLKTFDEK ELELIICGLG KIDVNDWKVN TRLKHCTPDS NIVKWFWKAV EFFDEERRAR LLQFVTGSSR VPLQGFKALQ GAAGPRLFTI HQIDACTNNL PKAHTCFNRI DIPPYESYEK LYEKLLTAIE ETCGFAVE
Greater than 90% as determined by SDS-PAGE.
64750
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers