missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ECI1 Protein

A cDNA sequence encoding the ECI1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11740-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15978198

  • 1740.00 € / 100µg
Estimated Shipment: 17-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1632
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSFGSQRVL VEPDAGAGVA VMKFKNPPVN SLSLEFLTEL VISLEKLEND KSFRGVILTS DRPGVFSAGL DLTEMCGRSP AHYAGYWKAV QELWLRLYQS NLVLVSAING ACPAGGCLVA LTCDYRILAD NPRYCIGLNE TQLGIIAPFW LKDTLENTIG HRAAERALQL GLLFPPAEAL QVGIVDQVVP EEQVQSTALS AIAQWMAIPD HARQLTKAMM RKATASRLVT QRDADVQNFV SFISKDSIQK SLQMYLERLK EEKG
Greater than 90.0% as determined by SDS-PAGE.
ECI1 Protein
Research Use Only
ECI1
Human
His
ECI1 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers