missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EDN2 Protein

Product Code. p-7149585
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 15918228

Brand: enQuireBio™ QP117461mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the EDN2 was constructed and used to recombinantly synthesize the protein.

Specifications

Name EDN2 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species Human
Protein Tag Untagged
Sequence MGSSHHHHHH SSGLVPRGSH MGSQAAATLE QPASSSHAQG THLRLRRCSC SSWLDKECVY FCHLDIIWVN TPEQTAPYGL GNPPRRRRRS LPRRCQCSSA RDPACATFCL RRPWTEAGAV PSRKSPADVF QTGKTGATTG ELLQRLRDIS TVKSLFAKRQ QEAMREPRST HSRWRKR
Buffer EDN2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol.
Purity or Quality Grade Greater than 85.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.