missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EDN3 Protein
A cDNA sequence encoding the EDN3 was constructed and used to recombinantly synthesize the protein.
433.00 € - 3320.00 €
Specifications
Gene ID (Entrez) | 1908 |
---|---|
Name | EDN3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | EDN3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15988228
|
enQuireBio™
QP11747-1MG |
1 mg |
433.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15998228
|
enQuireBio™
QP11747-5MG |
5 mg |
1935.00 €
5mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978228
|
enQuireBio™
QP11747-10MG |
10 mg |
3320.00 €
10mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
1908 | |
Research Use Only | |
EDN3 | |
Human | |
Untagged | |
The protein (1 mg/ml) was lyophilized with no additives. |
EDN3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
H-Cys-Thr-Cys-Phe-Thr-Tyr-Lys-Asp-Lys-Glu-Cys-Val-Tyr-Tyr-Cys-His-Leu-Asp-Ile-Ile-Trp-OH | |
Greater than 97.0% as determined by RP-HPLC. |