missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EED Protein
A cDNA sequence encoding the EED was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11748-1mg
Additional Details : Weight : 0.01000kg
Specifications
EED Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSEREVS TAPAGTDMPA AKKQKLSSDE NSNPDLSGDE NDDAVSIESG TNTERPDTPT NTPNAPGRKS WGKGKWKSKK CKYSFKCVNS LKEDHNQPLF GVQFNWHSKE GDPLVFATVG SNRVTLYECH SQGEIRLLQS YVDADADENF YTCAWTYDSN TSHPLLAVAG SRGIIRIINP ITMQCIKHYV GHGNAINELK FHPRDPNLLL SVSKDHALRL WNIQTDTLVA IFGGVEGHRD EVLSADYDLL GEKIMSCGMD HSLKLWRINS KRMMNAIKES YDYNPNKTNR PFISQKIHFP DFSTRDIHRN YVDCVRWLGD LILSKSCENA IVCWKPGKME DDIDKIKPSE SNVTILGRFD YSQCDIWYMR FSMDFWQKML ALGNQVGKLY VWDLEVEDPH KAKCTTLTHH KCGAAIRQTS FSRDSSILIA VCDDASIWRW DRLR | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The EED solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |