missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EED Protein

A cDNA sequence encoding the EED was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11748-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958238

  • 131.00 € / 5µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

EED Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSEREVS TAPAGTDMPA AKKQKLSSDE NSNPDLSGDE NDDAVSIESG TNTERPDTPT NTPNAPGRKS WGKGKWKSKK CKYSFKCVNS LKEDHNQPLF GVQFNWHSKE GDPLVFATVG SNRVTLYECH SQGEIRLLQS YVDADADENF YTCAWTYDSN TSHPLLAVAG SRGIIRIINP ITMQCIKHYV GHGNAINELK FHPRDPNLLL SVSKDHALRL WNIQTDTLVA IFGGVEGHRD EVLSADYDLL GEKIMSCGMD HSLKLWRINS KRMMNAIKES YDYNPNKTNR PFISQKIHFP DFSTRDIHRN YVDCVRWLGD LILSKSCENA IVCWKPGKME DDIDKIKPSE SNVTILGRFD YSQCDIWYMR FSMDFWQKML ALGNQVGKLY VWDLEVEDPH KAKCTTLTHH KCGAAIRQTS FSRDSSILIA VCDDASIWRW DRLR
Greater than 85% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The EED solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers