missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF4EBP2 Protein
A DNA sequence encoding the Homo sapiens (Human) EIF4EBP2, was expressed in the hosts and tags indicated.
202.00 € - 2645.00 €
Specifications
Accession Number | NP_004087.1 |
---|---|
Gene ID (Entrez) | 1979 |
Name | EIF4EBP2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969049
|
enQuireBio™
QP5973-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989049
|
enQuireBio™
QP5973-YE-1MG |
1 mg |
2645.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979049
|
enQuireBio™
QP5973-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15999049
|
enQuireBio™
QP5973-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15909059
|
enQuireBio™
QP5973-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15919059
|
enQuireBio™
QP5973-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004087.1 | |
EIF4EBP2 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1979 | |
Research Use Only | |
EIF4EBP2 | |
Recombinant Protein | |
Yeast | |
MSSSAGSGHQ PSQSRAIPTR TVAISDAAQL PHDYCTTPGG TLFSTTPGGT RIIYDRKFLL DRRNSPMAQT PPCHLPNIPG VTSPGTLIED SKVEVNNLNN LNNHDRKHAV GDDAQFEMDI | |
Greater than 90% as determined by SDS-PAGE. |