missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EIF4EBP2 Protein

A DNA sequence encoding the Homo sapiens (Human) EIF4EBP2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5973-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929049

  • 2285.00 € / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004087.1
EIF4EBP2 Protein
Research Use Only
EIF4EBP2
Recombinant Protein
E. coli
MSSSAGSGHQ PSQSRAIPTR TVAISDAAQL PHDYCTTPGG TLFSTTPGGT RIIYDRKFLL DRRNSPMAQT PPCHLPNIPG VTSPGTLIED SKVEVNNLNN LNNHDRKHAV GDDAQFEMDI
Greater than 90% as determined by SDS-PAGE.
1979
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers