missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ELAC1 Protein
A cDNA sequence encoding the ELAC1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | ELAC1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15908338
|
enQuireBio™
QP11777-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988328
|
enQuireBio™
QP11777-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15998328
|
enQuireBio™
QP11777-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ELAC1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ELAC1 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSMDVTF LGTGAAYPSP TRGASAVVLR CEGECWLFDC GEGTQTQLMK SQLKAGRITK IFITHLHGDH FFGLPGLLCT ISLQSGSMVS KQPIEIYGPV GLRDFIWRTM ELSHTELVFH YVVHELVPTA DQCPAEELKE FAHVNRADSP PKEEQGRTIL LDSEENSYLL FDDEQFVVKA FRLFHRIPSF GFSVVEKKRP GKLNAQKLKD LGVPPGPAYG KLKNGISVVL ENGVTISPQD VLKKPIVGRK ICILGDCSGV VGDGGVKLCF EADLLIHEAT LDDAQMDKAK EHGHSTPQMA ATFAKLCRAK RLVLTHFSQR YKPVALAREG ETDGIAELKK QAESVLDLQE VTLAEDFMVI SIPIKK | |
Greater than 80.0% as determined by SDS-PAGE. |