missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human ELFN2 Fc Chimera Protein
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human mGluR7 Protein (Catalog # 9530-UR ) is coated at 1 μg/mL, 100 μL/well, Recombinant Human ELFN2 Fc Chimera binds with an ED50 of 0.15-0.75 μg/mL.
Specifications
Specifications
| Accession Number | Q5R3F8.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 114794 |
| Molecular Weight (g/mol) | M.W.-observed: 97-112 kDa, under reducing conditions, M.W.-theroretical: 69 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human ELFN2 protein Human ELFN2 (Asp23-Tyr397) (N-terminus) Accession # Q5R3F8.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | dJ63G5.3, dJ63G5.3 (putative Leucine rich protein), ELFN2, extracellular leucine-rich repeat and fibronectin type III containing 2, extracellular leucine-rich repeat and fibronectin type III domain containing 2, extracellular leucine-rich repeat and fibronectin type III domain-containing protein 2, KIAA1904, leucine rich repeat containing 62, leucine-rich repeat and fibronectin type-III domain-containing protein 6, leucine-rich repeat-containing protein 62, LRRC62, PPP1R29, protein phosphatase 1, regulatory subunit 29 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?