missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EMC2 Protein
A cDNA sequence encoding the EMC2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11781-1mg
Additional Details : Weight : 0.01000kg
Specifications
EMC2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAKVSEL YDVTWEEMRD KMRKWREENS RNSEQIVEVG EELINEYASK LGDDIWIIYE QVMIAALDYG RDDLALFCLQ ELRRQFPGSH RVKRLTGMRF EAMERYDDAI QLYDRILQED PTNTAARKRK IAIRKAQGKN VEAIRELNEY LEQFVGDQEA WHELAELYIN EHDYAKAAFC LEELMMTNPH NHLYCQQYAE VKYTQGGLEN LELSRKYFAQ ALKLNNRNMR ALFGLYMSAS HIASNPKASA KTKKDNMKYA SWAASQINRA YQFAGRSKKE TKYSLKAVED MLETLQITQS | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The EMC2 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 2mM DTT and 40% glycerol. |