missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EML2 Protein
A DNA sequence encoding the Homo sapiens (Human) EML2, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001180197.1 |
---|---|
Gene ID (Entrez) | 24139 |
Name | EML2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15989119
|
enQuireBio™
QP5982-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15909129
|
enQuireBio™
QP5982-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999119
|
enQuireBio™
QP5982-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15919129
|
enQuireBio™
QP5982-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15929129
|
enQuireBio™
QP5982-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15939129
|
enQuireBio™
QP5982-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001180197.1 | |
EML2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
24139 | |
Research Use Only | |
EML2 | |
Recombinant Protein | |
E. coli | |
MSSFGAGKTK EVIFSVEDGS VKMFLRGRPV PMMIPDELAP TYSLDTRSEL PSCRLKLEWV YGYRGRDCRA NLYLLPTGEI VYFVASVAVL YSVEEQRQRH YLGHNDDIKC LAIHPDMVTI ATGQVAGTTK EGKPLPPHVR IWDSVSLSTL HVLGLGVFDR AVCCVGFSKS NGGNLLCAVD ESNDHMLSVW DWAKETKVVD VKCSNEAVLV ATFHPTDPTV LITCGKSHIY FWTLEGGSLS KRQGLFEKHE KPKYVLCVTF LEGGDVVTGD SGGNLYVWGK GGNRITQAVL GAHDGGVFGL CALRDGTLVS GGGRDRRVVL WGSDYSKLQE VEVPEDFGPV RTVAEGHGDT LYVGTTRNSI LQGSVHTGFS LLVQGHVEEL WGLATHPSRA QFVTCGQDKL VHLWSSDSHQ PLWSRIIE | |
Greater than 90% as determined by SDS-PAGE. |