missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Endoplasmic reticulum resident protein 29 Protein
A DNA sequence encoding the Homo sapiens (Human) Endoplasmic reticulum resident protein 29, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001029197.1 |
---|---|
Gene ID (Entrez) | 10961 |
Name | Endoplasmic reticulum resident protein 29 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044991
|
enQuireBio™
QP8504-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064991
|
enQuireBio™
QP8504-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054991
|
enQuireBio™
QP8504-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074991
|
enQuireBio™
QP8504-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084991
|
enQuireBio™
QP8504-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094991
|
enQuireBio™
QP8504-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001029197.1 | |
Endoplasmic reticulum resident protein 29 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10961 | |
Research Use Only | |
C12orf8 | |
Recombinant Protein | |
E. coli | |
PLDTVTFYKV IPKSKFVLVK FDTQYPYGEK QDEFKRLAEN SASSDDLLVA EVGISDYGDK LNMELSEKYK LDKESYPVFY LFRDGDFENP VPYTGAVKVG AIQRWLKGQG VYLGMPGCLP VYDALAGEFI RASGVEARQA LLKQGQDNLS SVKETQKKWA EQYLKIMGKI LDQGEDFPAS EMTRIARLIE KNKMSDGKKE ELQKSLNILT AF | |
Greater than 90% as determined by SDS-PAGE. |