missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ENTPD3 Protein

A cDNA sequence encoding the ENTPD3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11794-HIS-INSECT-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928398

  • 181.00 € / 5µg
Estimated Shipment: 20-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

956
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
ENTPD3
Human
His
ENTPD3 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
ENTPD3 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Insect
MGSSHHHHHH SSGLVPRGSH MGSQIHKQEV LPPGLKYGIV LDAGSSRTTV YVYQWPAEKE NNTGVVSQTF KCSVKGSGIS SYGNNPQDVP RAFEECMQKV KGQVPSHLHG STPIHLGATA GMRLLRLQNE TAANEVLESI QSYFKSQPFD FRGAQIISGQ EEGVYGWITA NYLMGNFLEK NLWHMWVHPH GVETTGALDL GGASTQISFV AGEKMDLNTS DIMQVSLYGY VYTLYTHSFQ CYGRNEAEKK FLAMLLQNSP TKNHLTNPCY PRDYSISFTM GHVFDSLCTV DQRPESYNPN DVITFEGTGD PSLCKEKVAS IFDFKACHDQ ETCSFDGVYQ PKIKGPFVAF AGFYYTASAL NLSGSFSLDT FNSSTWNFCS QNWSQLPLLL PKFDEVYARS YCFSANYIYH LFVNGYKFTE ETWPQIHFEK EVGNSSIAWS LGYMLSLTNQ IPAESPLIRL PIEPP
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers