missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ERO1L Protein

A cDNA sequence encoding the ERO1L was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11807-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15978438

  • 131.00 € / 5µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ERO1L Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSEEQPPET AAQRCFCQVS GYLDDCTCDV ETIDRFNNYR LFPRLQKLLE SDYFRYYKVN LKRPCPFWND ISQCGRRDCA VKPCQSDEVP DGIKSASYKY SEEANNLIEE CEQAERLGAV DESLSEETQK AVLQWTKHDD SSDNFCEADD IQSPEAEYVD LLLNPERYTG YKGPDAWKIW NVIYEENCFK PQTIKRPLNP LASGQGTSEE NTFYSWLEGL CVEKRAFYRL ISGLHASINV HLSARYLLQE TWLEKKWGHN ITEFQQRFDG ILTEGEGPRR LKNLYFLYLI ELRALSKVLP FFERPDFQLF TGNKIQDEEN KMLLLEILHE IKSFPLHFDE NSFFAGDKKE AHKLKEDFRL HFRNISRIMD CVGCFKCRLW GKLQTQGLGT ALKILFSEKL IANMPESGPS YEFHLTRQEI VSLFNAFGRI STSVKELENF RNLLQNIH
Greater than 90.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The ERO1L solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers