missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ETFDH Protein

A DNA sequence encoding the Homo sapiens (Human) ETFDH, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7564-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049430

  • 1665.00 € / 500µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001268666.1
ETFDH Protein
Research Use Only
ETFDH
Recombinant Protein
Yeast
SSTSTVPRIT THYTIYPRDK DKRWEGVNME RFAEEADVVI VGAGPAGLSA AVRLKQLAVA HEKDIRVCLV EKAAQIGAHT LSGACLDPGA FKELFPDWKE KGAPLNTPVT EDRFGILTEK YRIPVPILPG LPMNNHGNYI VRLGHLVSWM GEQAEALGVE VYPGYAAAEV LFHDDGSVKG IATNDVGIQK DGAPKATFER GLELHAKVTI FAEGCHGHLA KQLYKKFDLR ANCEPQTYGI GLKELWVIDE KNWKPGRVDH TVGWPLDRHT YGGSFLYHLN EGEPLVALGL VVGLDYQNPY LSPFREFQRW KHHPSIRPTL EGGKRIAYGA RALNEGGFQS IPKLTFPGGL LIGCSPGFMN VPKIKGTHTA MKSGILAAES IFNQLTSENL QSKTIGLHVT EYEDNLKNSW VWKELYSVRN IRPSCHGVLG VYGGMIYTGI FYWILRGMEP WTLKHKGSDF ERLKPAKDCT PIEYPKPDGQ ISFDLLSSVA LSGTNHEHDQ PAHLTLRDDS IPVNRNLSIY DGPEQRFCPA GVYEFVPVEQ GDGFRLQINA QNCVHCKTCD IKDPSQNINW VVPEGGGGPA YNGM
Greater than 90% as determined by SDS-PAGE.
2110
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers