missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Extracellular calcium-sensing receptor Protein

Product Code. p-7152378
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 15967589

Brand: enQuireBio™ QP5764ye10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Extracellular calcium-sensing receptor, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_000379.2
Gene ID (Entrez) 846
Name Extracellular calcium-sensing receptor Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol CASR
Protein Length Extracellular Domain
Product Type Recombinant Protein
Cross Reactivity Human
Species Yeast
Protein Tag His
Sequence YGPDQRAQKK GDIILGGLFP IHFGVAAKDQ DLKSRPESVE CIRYNFRGFR WLQAMIFAIE EINSSPALLP NLTLGYRIFD TCNTVSKALE ATLSFVAQNK IDSLNLDEFC NCSEHIPSTI AVVGATGSGV STAVANLLGL FYIPQVSYAS SSRLLSNKNQ FKSFLRTIPN DEHQATAMAD IIEYFRWNWV GTIAADDDYG RPGIEKFREE AEERDICIDF SELISQYSDE EEIQHVVEVI QNSTAKVIVV FSSGPDLEPL IKEIVRRNIT GKIWLASEAW ASSSLIAMPQ YFHVVGGTIG FALKAGQIPG FREFLKKVHP RKSVHNGFAK EFWEETFNCH LQEGAKGPLP VDTFLRGHEE SGDRFSNSST AFRPLCTGDE NISSVETPYI DYTHLRISYN VYLAVYSIAH ALQDIYTCLP GRGLFTNGSC ADIKKVEAWQ VLKHLRHLNF TNNMGEQVTF DECGDLVGNY SIINWHLSPE DGSIVFKEVG YYNVYAKKGE RLFINEEKIL WSGFSREVPF SNCSRDCLAG TRKGIIEGEP TCCFECVECP DGEYSDETDA SACNKCPDDF WSNENHTSCI AKEIEFLSWT EPF
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.